Assay Details
| Gene Symbol: LOC102136862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283283.1 | NP_001270212.1 | 1 - 2 | 99 | 94 | |
| XM_015448837.1 | XP_015304323.1 | 2 - 3 | 374 | 94 | ||
| GenBank mRNA | AB731598.1 | - | 1 - 2 | 81 | 94 | |
| FR719024.1 | - | 1 - 2 | 105 | 94 | ||
| FR719028.1 | - | 1 - 2 | 105 | 94 | ||
| HE573229.1 | - | 1 - 2 | 99 | 94 | ||
| HG007849.1 | - | 1 - 2 | 99 | 94 | ||
| HM579968.1 | - | 1 - 2 | 105 | 94 | ||
| HM579972.1 | - | 1 - 2 | 105 | 94 | ||
| HM579974.1 | - | 1 - 2 | 105 | 94 | ||
| HM579975.1 | - | 1 - 2 | 105 | 94 | ||
| HM580031.1 | - | 1 - 2 | 105 | 94 | ||
| LM654314.1 | - | 1 - 2 | 99 | 94 | ||
| LM654324.1 | - | 1 - 2 | 99 | 94 | ||
| LM654325.1 | - | 1 - 2 | 99 | 94 | ||
| LM654326.1 | - | 1 - 2 | 99 | 94 | ||
More Information
Set Membership: |
|

LOC102136862
Genomic Map