Assay Details
| Gene Symbol: LOC102136862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283283.1 | NP_001270212.1 | 2 - 3 | 346 | 117 | |
| XM_015448837.1 | XP_015304323.1 | 3 - 4 | 117 | |||
| GenBank mRNA | AB731597.1 | - | 2 - 3 | 328 | 117 | |
| AB731598.1 | - | 2 - 3 | 328 | 117 | ||
| AB764122.1 | - | 2 - 3 | 328 | 117 | ||
| AB764126.1 | - | 2 - 3 | 328 | 117 | ||
| AM943632.1 | - | 2 - 3 | 352 | 117 | ||
| AM943633.1 | - | 2 - 3 | 352 | 117 | ||
| EZ933315.1 | - | 2 - 3 | 346 | 117 | ||
| EZ933316.1 | - | 2 - 3 | 346 | 117 | ||
| FR719022.1 | - | 2 - 3 | 352 | 117 | ||
| FR719023.1 | - | 2 - 3 | 352 | 117 | ||
| FR719024.1 | - | 2 - 3 | 117 | |||
| FR719025.1 | - | 2 - 3 | 352 | 117 | ||
| FR719027.1 | - | 2 - 3 | 352 | 117 | ||
| FR719028.1 | - | 2 - 3 | 352 | 117 | ||
| FR719029.1 | - | 2 - 3 | 352 | 117 | ||
| HE573229.1 | - | 2 - 3 | 346 | 117 | ||
| HE573230.1 | - | 2 - 3 | 346 | 117 | ||
| HG007844.1 | - | 2 - 3 | 117 | |||
| HG007845.1 | - | 2 - 3 | 314 | 117 | ||
| HG007849.1 | - | 2 - 3 | 346 | 117 | ||
| HM579965.1 | - | 2 - 3 | 352 | 117 | ||
| HM579966.1 | - | 2 - 3 | 352 | 117 | ||
| HM579967.1 | - | 2 - 3 | 352 | 117 | ||
| HM579968.1 | - | 2 - 3 | 117 | |||
| HM579970.1 | - | 2 - 3 | 352 | 117 | ||
| HM579971.1 | - | 2 - 3 | 351 | 117 | ||
| HM579972.1 | - | 2 - 3 | 352 | 117 | ||
| HM579974.1 | - | 2 - 3 | 352 | 117 | ||
| HM579975.1 | - | 2 - 3 | 352 | 117 | ||
| HM580025.1 | - | 2 - 3 | 352 | 117 | ||
| HM580026.1 | - | 2 - 3 | 352 | 117 | ||
| HM580027.1 | - | 2 - 3 | 352 | 117 | ||
| HM580029.1 | - | 2 - 3 | 352 | 117 | ||
| HM580030.1 | - | 2 - 3 | 117 | |||
| HM580031.1 | - | 2 - 3 | 117 | |||
| LM654313.1 | - | 2 - 3 | 346 | 117 | ||
| LM654314.1 | - | 2 - 3 | 117 | |||
| LM654315.1 | - | 2 - 3 | 346 | 117 | ||
| LM654316.1 | - | 2 - 3 | 117 | |||
| LM654318.1 | - | 2 - 3 | 346 | 117 | ||
| LM654319.1 | - | 2 - 3 | 346 | 117 | ||
| LM654321.1 | - | 2 - 3 | 346 | 117 | ||
| LM654323.1 | - | 2 - 3 | 346 | 117 | ||
| LM654324.1 | - | 2 - 3 | 346 | 117 | ||
| LM654325.1 | - | 2 - 3 | 346 | 117 | ||
| LM654326.1 | - | 2 - 3 | 346 | 117 | ||
| LM654328.1 | - | 2 - 3 | 346 | 117 | ||
| LM654335.1 | - | 2 - 3 | 346 | 117 | ||
| LM654337.1 | - | 2 - 3 | 346 | 117 | ||
More Information
Set Membership: |
|

LOC102136862
Genomic Map