Assay Details
| Gene Symbol: LOC102136862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283283.1 | NP_001270212.1 | 3 - 4 | 628 | 104 | |
| XM_015448837.1 | XP_015304323.1 | 4 - 5 | 903 | 104 | ||
| GenBank mRNA | AB731598.1 | - | 3 - 4 | 610 | 104 | |
| AB764122.1 | - | 3 - 4 | 610 | 104 | ||
| AM943632.1 | - | 3 - 4 | 634 | 104 | ||
| AM943633.1 | - | 3 - 4 | 634 | 104 | ||
| EZ933316.1 | - | 3 - 4 | 628 | 104 | ||
| FR719025.1 | - | 3 - 4 | 634 | 104 | ||
| HE573229.1 | - | 3 - 4 | 628 | 104 | ||
| HG007849.1 | - | 3 - 4 | 628 | 104 | ||
| HM579967.1 | - | 3 - 4 | 634 | 104 | ||
| HM579974.1 | - | 3 - 4 | 634 | 104 | ||
| HM579975.1 | - | 3 - 4 | 634 | 104 | ||
| LM654320.1 | - | 3 - 4 | 628 | 104 | ||
| LM654327.1 | - | 3 - 4 | 628 | 104 | ||
| LM654335.1 | - | 3 - 4 | 628 | 104 | ||
More Information
Set Membership: |
|

LOC102136862
Genomic Map