Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284165.1 | NP_001271094.1 | 3 - 4 | 600 | 86 | |
| XM_005589764.2 | XP_005589821.1 | 4 - 5 | 881 | 86 | ||
| GenBank mRNA | AB174530.1 | - | 3 - 4 | 600 | 86 | |
More Information
Set Membership: |
|

TMEM143
Genomic Map