Assay Details
| Gene Symbol: PRDX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284928.1 | NP_001271857.1 | 3 - 4 | 327 | 99 | |
| GenBank mRNA | AB174701.1 | - | 3 - 4 | 327 | 99 | |
More Information
Set Membership: |
|

PRDX3
Genomic Map