Assay Details
| Gene Symbol: BBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285040.1 | NP_001271969.1 | 3 - 4 | 672 | 77 | |
| XM_005578360.2 | XP_005578417.1 | 3 - 4 | 385 | 77 | ||
| XM_015434088.1 | XP_015289574.1 | 4 - 5 | 843 | 77 | ||
| GenBank mRNA | AB171325.1 | - | 3 - 4 | 672 | 77 | |
More Information
Set Membership: |
|

BBOX1
Genomic Map