Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283723.1 | NP_001270652.1 | 3 - 4 | 615 | 72 | |
| XM_005570174.2 | XP_005570231.1 | 3 - 4 | 832 | 72 | ||
| XM_015430178.1 | XP_015285664.1 | 3 - 4 | 634 | 72 | ||
| XM_015430179.1 | XP_015285665.1 | 3 - 4 | 540 | 72 | ||
| GenBank mRNA | AB060888.1 | - | 3 - 4 | 615 | 72 | |
More Information
Set Membership: |
|

MANSC1
Genomic Map