Assay Details
| Gene Symbol: C11H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284593.1 | NP_001271522.1 | 2 - 3 | 456 | 107 | |
| XM_015430722.1 | XP_015286208.1 | 2 - 3 | 533 | 107 | ||
| GenBank mRNA | AB168147.1 | - | 2 - 3 | 456 | 107 | |
More Information
Set Membership: |
|

C11H12orf56
Genomic Map