Assay Details
| Gene Symbol: C11H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284593.1 | NP_001271522.1 | 4 - 5 | 921 | 103 | |
| XM_015430721.1 | XP_015286207.1 | 4 - 5 | 959 | 103 | ||
| XM_015430722.1 | XP_015286208.1 | 4 - 5 | 998 | 103 | ||
| GenBank mRNA | AB168147.1 | - | 4 - 5 | 921 | 103 | |
More Information
Set Membership: |
|

C11H12orf56
Genomic Map