Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285060.2 | NP_001271989.2 | 3 - 4 | 695 | 94 | |
| XM_005549983.2 | XP_005550040.1 | 3 - 4 | 694 | 94 | ||
| GenBank mRNA | AB170507.1 | - | 3 - 4 | 95 | ||
More Information
Set Membership: |
|

CCDC126
Genomic Map