Assay Details
| Gene Symbol: SLC22A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284728.1 | NP_001271657.1 | 4 - 5 | 815 | 60 | |
| XM_015445292.1 | XP_015300778.1 | 4 - 5 | 974 | 60 | ||
| XM_015445293.1 | XP_015300779.1 | 4 - 5 | 1683 | 60 | ||
| XM_015445294.1 | XP_015300780.1 | 4 - 5 | 1334 | 60 | ||
| XM_015445295.1 | XP_015300781.1 | 6 - 7 | 993 | 60 | ||
| XM_015445296.1 | XP_015300782.1 | 5 - 6 | 1413 | 60 | ||
| XM_015445297.1 | XP_015300783.1 | 6 - 7 | 980 | 60 | ||
| XM_015445298.1 | XP_015300784.1 | 4 - 5 | 1241 | 60 | ||
| GenBank mRNA | AB168855.1 | - | 4 - 5 | 815 | 60 | |
More Information
Set Membership: |
|

SLC22A14
Genomic Map