Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285126.1 | NP_001272055.1 | 5 - 6 | 476 | 105 | |
| XM_005576632.2 | XP_005576689.1 | 6 - 7 | 903 | 105 | ||
| XM_005576634.2 | XP_005576691.1 | 5 - 6 | 1199 | 105 | ||
| XM_015433422.1 | XP_015288908.1 | 5 - 6 | 852 | 105 | ||
| XM_015433423.1 | XP_015288909.1 | 4 - 5 | 783 | 105 | ||
| GenBank mRNA | AB171815.1 | - | 5 - 6 | 476 | 105 | |
More Information
Set Membership: |
|

COLEC11
Genomic Map