Assay Details
| Gene Symbol: TUBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285304.1 | NP_001272233.1 | 3 - 4 | 556 | 96 | |
| XM_005583880.2 | XP_005583937.1 | 3 - 4 | 560 | 96 | ||
| XM_005583882.2 | XP_005583939.1 | 3 - 4 | 560 | 96 | ||
| XM_015437804.1 | XP_015293290.1 | 3 - 4 | 560 | 96 | ||
| XM_015437805.1 | XP_015293291.1 | 2 - 3 | 405 | 96 | ||
| GenBank mRNA | AB168725.1 | - | 3 - 4 | 556 | 96 | |
More Information
Set Membership: |
|

TUBD1
Genomic Map