Assay Details
| Gene Symbol: CALM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284121.1 | NP_001271050.1 | 4 - 5 | 404 | 80 | |
| GenBank mRNA | AB169886.1 | - | 4 - 5 | 404 | 80 | |
| AB172317.1 | - | 4 - 5 | 80 | |||
More Information
Set Membership: |
|

CALM3
Genomic Map