Assay Details
| Gene Symbol: XRCC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284708.1 | NP_001271637.1 | 5 - 6 | 729 | 61 | |
| XM_005567141.2 | XP_005567198.1 | 5 - 6 | 801 | 61 | ||
| XM_015456936.1 | XP_015312422.1 | 5 - 6 | 617 | 61 | ||
| GenBank mRNA | AB168631.1 | - | 5 - 6 | 729 | 61 | |
| AB169317.1 | - | 5 - 6 | 649 | 61 | ||
| AB170097.1 | - | 5 - 6 | 649 | 61 | ||
More Information
Set Membership: |
|

XRCC6
Genomic Map