Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284830.1 | NP_001271759.1 | 2 - 3 | 744 | 87 | |
| GenBank mRNA | AB170735.1 | - | 2 - 3 | 778 | 87 | |
| AB170987.1 | - | 2 - 3 | 744 | 87 | ||
More Information
Set Membership: |
|

CAMLG
Genomic Map