Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284960.1 | NP_001271889.1 | 2 - 3 | 418 | 91 | |
| XM_005561059.2 | XP_005561116.1 | 2 - 3 | 452 | 91 | ||
| XM_015453219.1 | XP_015308705.1 | 2 - 3 | 702 | 91 | ||
| GenBank mRNA | AB170079.1 | - | 2 - 3 | 418 | 91 | |
More Information
Set Membership: |
|

CFL2
Genomic Map