Assay Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283405.1 | NP_001270334.1 | 4 - 5 | 427 | 57 | |
| XM_015436100.1 | XP_015291586.1 | 4 - 5 | 689 | 57 | ||
| XM_015436101.1 | XP_015291587.1 | 3 - 4 | 682 | 57 | ||
| GenBank mRNA | AB172284.1 | - | 4 - 5 | 427 | 57 | |
More Information
Set Membership: |
|

COQ4
Genomic Map