Assay Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285026.1 | NP_001271955.1 | 4 - 5 | 1404 | 90 | |
| XM_005543781.2 | XP_005543838.1 | 5 - 6 | 5490 | 90 | ||
| XM_005543782.2 | XP_005543839.1 | 4 - 5 | 5510 | 90 | ||
| XM_005543783.2 | XP_005543840.1 | 4 - 5 | 5418 | 90 | ||
| XM_005543784.2 | XP_005543841.1 | 5 - 6 | 5425 | 90 | ||
| XM_005543785.2 | XP_005543842.1 | 5 - 6 | 5491 | 90 | ||
| XM_005543786.2 | XP_005543843.1 | 5 - 6 | 5489 | 90 | ||
| XM_005543789.2 | XP_005543846.1 | 6 - 7 | 5353 | 90 | ||
| XM_005543790.2 | XP_005543847.1 | 6 - 7 | 5380 | 90 | ||
| XM_015451487.1 | XP_015306973.1 | 4 - 5 | 1887 | 90 | ||
| XM_015451491.1 | XP_015306977.1 | 3 - 4 | 1972 | 90 | ||
| XM_015451495.1 | XP_015306981.1 | 6 - 7 | 5315 | 90 | ||
| GenBank mRNA | AB179349.1 | - | 4 - 5 | 1404 | 90 | |
More Information
Set Membership: |
|

SLFNL1
Genomic Map