Assay Details
| Gene Symbol: SERINC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284660.1 | NP_001271589.1 | 9 - 10 | 1064 | 80 | |
| XM_015451395.1 | XP_015306881.1 | 7 - 8 | 1009 | 80 | ||
| GenBank mRNA | AB169164.1 | - | 9 - 10 | 1064 | 80 | |
More Information
Set Membership: |
|

SERINC5
Genomic Map