Assay Details
| Gene Symbol: MED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284739.1 | NP_001271668.1 | 6 - 7 | 679 | 62 | |
| XM_015438962.1 | XP_015294448.1 | 6 - 7 | 698 | 62 | ||
| GenBank mRNA | AB174170.1 | - | 6 - 7 | 679 | 62 | |
More Information
Set Membership: |
|

MED4
Genomic Map