Assay Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285150.1 | NP_001272079.1 | 1 - 2 | 243 | 80 | |
| GenBank mRNA | AB169830.1 | - | 1 - 2 | 243 | 80 | |
More Information
Set Membership: |
|

ARL6IP5
Genomic Map