Assay Details
| Gene Symbol: TUBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283445.1 | NP_001270374.1 | 3 - 4 | 388 | 105 | |
| GenBank mRNA | AB125205.1 | - | 3 - 4 | 332 | 105 | |
| AB169662.1 | - | 3 - 4 | 388 | 105 | ||
More Information
Set Membership: |
|

TUBB3
Genomic Map