Assay Details
| Gene Symbol: PPP1R2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283605.1 | NP_001270534.1 | 1 | 620 | 81 | |
| GenBank mRNA | AB169376.1 | - | 1 | 620 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R2C| Gene Symbol: PPP1R2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283605.1 | NP_001270534.1 | 1 | 620 | 81 | |
| GenBank mRNA | AB169376.1 | - | 1 | 620 | 81 | |
Set Membership: |
|