Assay Details
| Gene Symbol: AAGAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284079.1 | NP_001271008.1 | 9 - 10 | 876 | 64 | |
| XM_005559881.2 | XP_005559938.1 | 9 - 10 | 1228 | 64 | ||
| XM_005559882.2 | XP_005559939.1 | 9 - 10 | 1122 | 64 | ||
| XM_015452436.1 | XP_015307922.1 | 9 - 10 | 1225 | 64 | ||
| GenBank mRNA | AB168215.1 | - | 9 - 10 | 876 | 64 | |
More Information
Set Membership: |
|

AAGAB
Genomic Map