Assay Details
| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284643.1 | NP_001271572.1 | 1 - 2 | 257 | 68 | |
| XM_015443747.1 | XP_015299233.1 | 1 - 2 | 404 | 68 | ||
| XM_015443748.1 | XP_015299234.1 | 1 - 2 | 302 | 68 | ||
| GenBank mRNA | AB170419.1 | - | 1 - 2 | 257 | 68 | |
More Information
Set Membership: |
|

SNX12
Genomic Map