Assay Details
| Gene Symbol: WDR41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283117.1 | NP_001270046.1 | 6 - 7 | 453 | 84 | |
| XM_015451398.1 | XP_015306884.1 | 5 - 6 | 756 | 84 | ||
| XM_015451399.1 | XP_015306885.1 | 5 - 6 | 756 | 84 | ||
| XM_015451400.1 | XP_015306886.1 | 5 - 6 | 756 | 84 | ||
| XM_015451401.1 | XP_015306887.1 | 6 - 7 | 574 | 84 | ||
| XM_015451402.1 | XP_015306888.1 | 5 - 6 | 773 | 84 | ||
| XM_015451403.1 | XP_015306889.1 | 5 - 6 | 375 | 84 | ||
| XM_015451404.1 | XP_015306890.1 | 5 - 6 | 773 | 84 | ||
| GenBank mRNA | AB170095.1 | - | 6 - 7 | 453 | 84 | |
More Information
Set Membership: |
|

WDR41
Genomic Map