Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283233.1 | NP_001270162.1 | 4 - 5 | 616 | 94 | |
| XM_005546054.1 | XP_005546111.1 | 4 - 5 | 678 | 94 | ||
| XM_005546056.1 | XP_005546113.1 | 4 - 5 | 678 | 94 | ||
| GenBank mRNA | AB169787.1 | - | 4 - 5 | 616 | 94 | |
More Information
Set Membership: |
|

GYG1
Genomic Map