Assay Details
| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283180.1 | NP_001270109.1 | 8 - 9 | 1351 | 61 | |
| XM_015445343.1 | XP_015300829.1 | 5 - 6 | 730 | 61 | ||
| GenBank mRNA | AB070185.1 | - | 8 - 9 | 1351 | 61 | |
More Information
Set Membership: |
|

ZPLD1
Genomic Map