Assay Details
| Gene Symbol: SRD5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283467.1 | NP_001270396.1 | 1 - 2 | 294 | 61 | |
| GenBank mRNA | S77165.1 | - | 1 - 2 | 294 | 61 | |
More Information
Set Membership: |
|

SRD5A2
Genomic Map