Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284580.1 | NP_001271509.1 | 3 - 4 | 356 | 80 | |
| XM_005590349.1 | XP_005590406.1 | 3 - 4 | 803 | 80 | ||
| GenBank mRNA | AJ278288.1 | - | 3 - 4 | 356 | 80 | |
More Information
Set Membership: |
|

NCR1
Genomic Map