Assay Details
| Gene Symbol: SPATA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287318.1 | NP_001274247.1 | 6 - 7 | 1189 | 67 | |
| XM_005561973.2 | XP_005562030.1 | 6 - 7 | 1252 | 67 | ||
| XM_005561974.2 | XP_005562031.1 | 6 - 7 | 1252 | 67 | ||
| XM_005561975.2 | XP_005562032.1 | 5 - 6 | 1156 | 67 | ||
| XM_005561977.2 | XP_005562034.1 | 7 - 8 | 1138 | 67 | ||
| XM_015453254.1 | XP_015308740.1 | 6 - 7 | 1170 | 67 | ||
| XM_015453255.1 | XP_015308741.1 | 6 - 7 | 1170 | 67 | ||
| XM_015453256.1 | XP_015308742.1 | 5 - 6 | 1109 | 67 | ||
| XM_015453257.1 | XP_015308743.1 | 7 - 8 | 1095 | 67 | ||
| GenBank mRNA | AB070123.1 | - | 6 - 7 | 1189 | 67 | |
| AB168761.1 | - | 5 - 6 | 511 | 67 | ||
More Information
Set Membership: |
|

SPATA7
Genomic Map