Assay Details
| Gene Symbol: C4H6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287704.1 | NP_001274633.1 | 1 | 720 | 82 | |
| XM_015448387.1 | XP_015303873.1 | 2 | 545 | 82 | ||
| GenBank mRNA | AB056416.1 | - | 1 | 720 | 82 | |
More Information
Set Membership: |
|

C4H6orf120
Genomic Map