Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283773.1 | NP_001270702.1 | 5 - 6 | 691 | 90 | |
| XM_015443720.1 | XP_015299206.1 | 5 - 6 | 760 | 90 | ||
| XM_015443721.1 | XP_015299207.1 | 5 - 6 | 760 | 90 | ||
| XM_015443722.1 | XP_015299208.1 | 5 - 6 | 760 | 90 | ||
| XM_015443723.1 | XP_015299209.1 | 5 - 6 | 760 | 90 | ||
| GenBank mRNA | AB168869.1 | - | 5 - 6 | 691 | 90 | |
More Information
Set Membership: |
|

RBMX
Genomic Map