Assay Details
| Gene Symbol: GSTM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285257.1 | NP_001272186.1 | 7 - 8 | 596 | 97 | |
| GenBank mRNA | AB169530.1 | - | 7 - 8 | 626 | 97 | |
| AB220566.1 | - | 7 - 8 | 596 | 97 | ||
| EF104257.1 | - | 7 - 8 | 576 | 97 | ||
More Information
Set Membership: |
|

GSTM5
Genomic Map