Assay Details
| Gene Symbol: MAD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283265.1 | NP_001270194.1 | 5 | 2018 | 95 | |
| GenBank mRNA | AB168907.1 | - | 5 | 2018 | 95 | |
More Information
Set Membership: |
|

MAD2L1
Genomic Map