Assay Details
| Gene Symbol: UBE2N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284878.1 | NP_001271807.1 | 4 | 1617 | 101 | |
| GenBank mRNA | AB169913.1 | - | 4 | 1617 | 101 | |
More Information
Set Membership: |
|

UBE2N
Genomic Map