Assay Details
| Gene Symbol: PNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284017.1 | NP_001270946.1 | 7 | 2220 | 105 | |
| GenBank mRNA | AB174453.1 | - | 7 | 2220 | 105 | |
More Information
Set Membership: |
|

PNO1
Genomic Map