Assay Details
| Gene Symbol: PRPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284579.1 | NP_001271508.1 | 7 | 1919 | 163 | |
| GenBank mRNA | AB169801.1 | - | 7 | 1919 | 163 | |
More Information
Set Membership: |
|

PRPS1
Genomic Map