Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302658.1 | NP_001289587.1 | 2 - 3 | 508 | 84 | |
| XM_015451946.1 | XP_015307432.1 | 2 - 3 | 532 | 84 | ||
More Information
Set Membership: |
|

SLAMF9
Genomic Map