Assay Details
| Gene Symbol: GML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564195.2 | XP_005564252.1 | 2 - 3 | 159 | 63 | |
| XM_005564196.2 | XP_005564253.1 | 2 - 3 | 116 | 63 | ||
More Information
Set Membership: |
|

GML
Genomic Map