Assay Details
| Gene Symbol: RAD51AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576694.2 | XP_005576751.2 | 2 - 3 | 3410 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAD51AP2| Gene Symbol: RAD51AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576694.2 | XP_005576751.2 | 2 - 3 | 3410 | 83 | |
Set Membership: |
|