Assay Details
Gene Symbol: GFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589900.2 | XP_005589957.1 | 4 - 5 | 1997 | 98 | |
XM_015441134.1 | XP_015296620.1 | 4 - 5 | 2254 | 98 | ||
XM_015441135.1 | XP_015296621.1 | 3 - 4 | 2046 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
