Assay Details
| Gene Symbol: LOC102124147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590008.2 | XP_005590065.1 | 5 - 6 | 529 | 87 | |
| XM_015441358.1 | XP_015296844.1 | 29 - 30 | 3866 | 87 | ||
| XM_015441359.1 | XP_015296845.1 | 3 - 4 | 398 | 87 | ||
| XM_015441360.1 | XP_015296846.1 | 2 - 3 | 218 | 87 | ||
More Information
Set Membership: |
|

LOC102124147
Genomic Map