Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447126.1 | XP_015302612.1 | 3 - 4 | 849 | 104 | |
| XM_015447128.1 | XP_015302614.1 | 3 - 4 | 909 | 104 | ||
| XM_015447129.1 | XP_015302615.1 | 3 - 4 | 849 | 104 | ||
More Information
Set Membership: |
|

TRIM4
Genomic Map