Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568545.2 | XP_005568602.1 | 3 - 4 | 986 | 94 | |
| XM_005568546.2 | XP_005568603.1 | 3 - 4 | 1022 | 94 | ||
More Information
Set Membership: |
|

SIRPB2
Genomic Map