Assay Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570599.2 | XP_005570656.1 | 6 - 7 | 1707 | 65 | |
| XM_005570601.2 | XP_005570658.1 | 6 - 7 | 1133 | 65 | ||
| XM_005570602.2 | XP_005570659.1 | 6 - 7 | 1072 | 65 | ||
| XM_005570603.2 | XP_005570660.1 | 6 - 7 | 869 | 65 | ||
| XM_005570604.2 | XP_005570661.1 | 6 - 7 | 1069 | 65 | ||
| XM_015430735.1 | XP_015286221.1 | 7 - 8 | 1252 | 65 | ||
More Information
Set Membership: |
|

PRICKLE1
Genomic Map