Assay Details
| Gene Symbol: INSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588399.2 | XP_005588456.1 | 1 - 2 | 255 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSL3| Gene Symbol: INSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588399.2 | XP_005588456.1 | 1 - 2 | 255 | 80 | |
Set Membership: |
|