Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558232.2 | XP_005558289.1 | 2 - 3 | 961 | 82 | |
| XM_015452124.1 | XP_015307610.1 | 2 - 3 | 961 | 82 | ||
More Information
Set Membership: |
|

SLC26A2
Genomic Map