Assay Details
| Gene Symbol: SEC14L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568097.2 | XP_005568154.1 | 5 - 6 | 490 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC14L6| Gene Symbol: SEC14L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568097.2 | XP_005568154.1 | 5 - 6 | 490 | 79 | |
Set Membership: |
|